Sodium hexanitritocobaltate(III) ACS

Supplier: BeanTown Chemical

Synonyms: Sodium cobaltnitrite, Trisodium hexanitritocobaltate

225535-100G 225535-500G
BT225535-100GEA 106.45 USD
BT225535-100G BT225535-500G
Sodium hexanitritocobaltate(III) ACS
Sodium hexanitritocobaltate(III)

CAS: 13600-98-1; EC No: 237-077-7; MDL No: MFCD00003512 UN No: UN1479; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: Na3Co(NO2)6; MW: 403.94 Melting Point: 220° (decomposes)


  • Powder

Formula: Na₃Co(NO₂)₆
MW: 403.94 g/mol
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00003512
CAS Number: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR