3-Chloroaniline ≥99%
Supplier: BeanTown Chemical
225525-100G
225525-500G
BT225525-100GEA
30
USD
BT225525-100G
BT225525-500G
3-Chloroaniline ≥99%
3-Chloroaniline
CAS: 108-42-9; EC No: 203-581-0; MDL No: MFCD00007765; RTECS: BX0350000
UN No: UN2019; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C6H6ClN; MW: 127.57
Melting Point: -10°; Boiling Point: 95-96°/11 mmHg; Flash point: 118°C (244°F)
Density (g/mL): 1.206; Refractive Index: 1.594
- RTECS: BX0350000
- Refractive index: 1.594
- Liquid
Formula:
C₆H₆ClN MW: 127.57 g/mol Boiling Pt: 95…96 °C/11 mmHg Melting Pt: –10 °C Density: 1.206 g/mL Flash Pt: 118 °C (244 °F) |
MDL Number:
MFCD00007765 CAS Number: 108-42-9 EINECS: 203-581-0 UN: 2019 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...