Silver ≥99.999% (trace metals basis), powder -120 mesh

Supplier: BeanTown Chemical

225505-25G 225505-5G
BT225505-25GEA 444 USD
BT225505-25G BT225505-5G
Silver ≥99.999% (trace metals basis), powder -120 mesh
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • RTECS: VW3500000
  • Powder

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
ADR: 9,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR