(R)-(-)-4-Penten-2-ol ≥95%

Supplier: BeanTown Chemical

Synonyms: (R)-(-)-1-Penten-4-ol, (R)-2-Hydroxypent-4-ene, (R)-(-)-Allyl methyl carbinol

225470-1G 225470-5G
BT225470-1GEA 153 USD
BT225470-1G BT225470-5G
(R)-(-)-4-Penten-2-ol ≥95%
(R)-(-)-4-Penten-2-ol

CAS: 64584-92-5; MDL No: MFCD03701536 UN No: UN1987; Haz Class: 3; Packing Group: III Liquid; Linear Formula: H2C=CHCH2CH(OH)CH3; Molecular Formula: C5H10O; MW: 86.13 Boiling Point: 115-116°; Flash point: 25°C (77°F) Density (g/mL): 0.837; Refractive Index: 1.424; Optical Rotation: [α]20/D −5.05°, neat


  • Optical rotation: [α]20/D −5.05°, neat
  • Refractive index: 1.424
  • Liquid

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 115…116 °C
Density: 0.837 g/mL
Flash Pt: 25 °C (77 °F)
MDL Number: MFCD03701536
CAS Number: 64584-92-5
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR