Cobalt(II) fluoride, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
Cobalt difluoride, Cobaltous fluoride
225465-100G
225465-25G
BT225465-100GEA
208
USD
BT225465-100G
BT225465-25G
Cobalt(II) fluoride, anhydrous ≥98%
Cobalt(II) fluoride
CAS: 10026-17-2; EC No: 233-061-9; MDL No: MFCD00010941; RTECS: GG0770000
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Powder; Molecular Formula: CoF2; MW: 96.93
Density (g/mL): 4.43
Hygroscopic
- RTECS: GG0770000
- Hygroscopic
- Powder
Formula:
CoF₂ MW: 96.93 g/mol Density: 4.43 g/mL |
MDL Number:
MFCD00010941 CAS Number: 10026-17-2 EINECS: 233-061-9 UN: 2923 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...