Tin(II) chloride, anhydrous ≥99%
Supplier: BeanTown Chemical
CAS: 7772-99-8; EC No: 231-868-0; MDL No: MFCD00011241; RTECS: XP8700000
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Linear Formula: SnCl2; MW: 189.62
Melting Point: 246°; Boiling Point: 652°
Density (g/mL): 3.95
Air Sensitive, Moisture Sensitive
- RTECS: XP8700000
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
SnCl₂ Boiling Pt: 652 °C Melting Pt: 246 °C Density: 3.95 g/mL |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 EINECS: 231-868-0 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...