Dicopper selenide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Copper(I) selenide

225410-10G 225410-50G
BT225410-10GEA 86 USD
BT225410-10G BT225410-50G
Dicopper selenide ≥99.5% (trace metals basis)
Dicopper selenide

CAS: 20405-64-5; EC No: 243-796-7; MDL No: MFCD00049908 UN No: UN3077; Haz Class: 9; Packing Group: III 2mm & Down Pieces; Molecular Formula: Cu2Se; MW: 206.05 Melting Point: 1113° Density (g/mL): 6.84


  • 2mm & Down Pieces

Formula: Cu₂Se
MW: 206.05 g/mol
Melting Pt: 1113 °C
Density: 6.84 g/mL
MDL Number: MFCD00049908
CAS Number: 20405-64-5
EINECS: 243-796-7
UN: 3077
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR