N,N-Dimethylacrylamide ≥99.5% stabilized

Supplier: BeanTown Chemical

225390-1KG 225390-250G 225390-50G
BT225390-1KGEA 446 USD
BT225390-1KG BT225390-250G BT225390-50G
N,N-Dimethylacrylamide ≥99.5% stabilized
N,N-Dimethylacrylamide

CAS: 2680-03-7; EC No: 220-237-5; MDL No: MFCD00008626; RTECS: AU3230000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: CH2=CHCON(CH3)2; Molecular Formula: C5H9NO; MW: 99.13 Melting Point: -40°; Boiling Point: 80-81°/20 mmHg; Flash point: 70°C (158°F) Density (g/mL): 0.962; Refractive Index: 1.473 Light Sensitive


  • RTECS: AU3230000
  • Refractive index: 1.473
  • Light Sensitive
  • Liquid

Formula: C₅H₉NO
Boiling Pt: 80…81 °C/20 mmHg
Melting Pt: –40 °C
Density: 0.962 g/mL
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00008626
CAS Number: 2680-03-7
EINECS: 220-237-5
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR