N,N-Dimethyl-p-toluidine ≥99%

Supplier: BeanTown Chemical

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

225385-100G 225385-2.5KG 225385-500G
BT225385-100GEA 29.34 USD
BT225385-100G BT225385-2.5KG BT225385-500G
N,N-Dimethyl-p-toluidine ≥99%
N,N-Dimethyl-p-toluidine

CAS: 99-97-8; EC No: 202-805-4; MDL No: MFCD00008316; RTECS: XU5803000 UN No: UN1708; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: CH3C6H4N(CH3)2; Molecular Formula: C9H13N; MW: 135.21 Melting Point: -25°; Boiling Point: 210-211°; Flash point: 83°C (181°F) Density (g/mL): 0.937; Refractive Index: 1.546


  • RTECS: XU5803000
  • Refractive index: 1.546
  • Liquid

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 210…211 °C
Melting Pt: –25 °C
Density: 0.937 g/mL
Flash Pt: 83 °C (181 °F)
MDL Number: MFCD00008316
CAS Number: 99-97-8
EINECS: 202-805-4
UN: 1708
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR