1-Pentene ≥97%

Supplier: BeanTown Chemical

Synonyms: alpha-n-Amylene, Propylethylene

225365-100G 225365-25G
BT225365-100GEA 180.62 USD
BT225365-100G BT225365-25G
1-Pentene ≥97%
1-Pentene

CAS: 109-67-1; EC No: 203-694-5; MDL No: MFCD00003567; RTECS: SB2179000 UN No: UN1108; Haz Class: 3; Packing Group: I Liquid; Linear Formula: CH3(CH2)2CH=CH2; Molecular Formula: C5H10 ; MW: 70.13 Boiling Point: 30-31°; Flash point: -51°C (-60°F) Density (g/mL): 0.641; Refractive Index: 1.371


  • RTECS: SB2179000
  • Refractive index: 1.371
  • Liquid

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 30…31 °C
Density: 0.641 g/mL
Flash Pt: –51 °C (–60 °F)
MDL Number: MFCD00003567
CAS Number: 109-67-1
EINECS: 203-694-5
UN: 1108
ADR: 3,I

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR