1,4-Dioxane ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

225360-1L 225360-500ML
BT225360-1LEA 92.2 USD
BT225360-1L BT225360-500ML
1,4-Dioxane ≥99.0% ACS
1,4-Dioxane

CAS: 123-91-1; EC No: 204-661-8; MDL No: MFCD00006571; RTECS: JG8225000 UN No: UN1165; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H8O2; MW: 88.11 Melting Point: 10-12°; Boiling Point: 100-102°; Flash point: 12°C (54°F) Density (g/mL): 1.034; Refractive Index: 1.422 Hygroscopic


  • RTECS: JG8225000
  • Refractive index: 1.422
  • Hygroscopic
  • Liquid

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1
Melting Pt: 11.8
Density: 1.034 g/mL
Flash Pt: 12 °C (54 °F)
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR