N-Methyl-4-nitroaniline ≥97%
Supplier: BeanTown Chemical
225260-100G
225260-25G
225260-5G
BT225260-100GEA
230
USD
BT225260-100G
BT225260-25G
BT225260-5G
N-Methyl-4-nitroaniline ≥97%
N-Methyl-4-nitroaniline
CAS: 100-15-2; EC No: 202-823-2; MDL No: MFCD00007305
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: NO2C6H4NHCH3; Molecular Formula: C7H8N2O2 ; MW: 152.15
Melting Point: 149-151°
- Powder
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 149…151 °C |
MDL Number:
MFCD00007305 CAS Number: 100-15-2 EINECS: 202-823-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...