Orthophosphoric acid ≥85% (w/w) in water ACS
Supplier: BeanTown Chemical
Synonyms:
Phosphoric acid
225220-1KG
225220-250G
BT225220-1KGEA
55.94
USD
BT225220-1KG
BT225220-250G
Orthophosphoric acid ≥85% (w/w) in water ACS
Orthophosphoric acid
CAS: 7664-38-2; EC No: 231-633-2; MDL No: MFCD00011340; RTECS: TB6300000
UN No: UN1805; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: H3PO4; MW: 98.00
Melting Point: 19-21°
Density (g/mL): 1.685; Refractive Index: 1.431
- RTECS: TB6300000
- Refractive index: 1.431
- Liquid
Formula:
H₃PO₄ MW: 98 g/mol Boiling Pt: 158 Melting Pt: 21 Density: 1.685 g/mL |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8 III Merck Index: 14,07344 |
Specification Test Results
Assay | 85-87% |
Color (APHA) | ≤10 |
Insoluble matter | ≤0.001% |
Chloride (Cl) | ≤3ppm |
Sulfate (SO4) | ≤0.003% |
Nitrate (NO3) | ≤5ppm |
Volatile acids (as CH3COOH) | ≤0.001% |
Antimony (Sb) | ≤0.002% |
Calcium (Ca) | ≤0.002% |
Magnesium (Mg) | ≤0.002% |
Potassium (K) | ≤0.005% |
Sodium (Na) | ≤0.025% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.2ppm |
Arsenic (As) | ≤1ppm |
Manganese (Mn) | ≤0.5ppm |
Reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...