Copper(I) bromide dimethyl sulfide complex

Supplier: BeanTown Chemical

225215-25G 225215-5G
BT225215-25GEA 205 USD
BT225215-25G BT225215-5G
Copper(I) bromide dimethyl sulfide complex
Copper (I) bromide dimethyl sulphide complex

CAS: 54678-23-8; EC No: 259-292-5; MDL No: MFCD00043295 UN No: UN3335; Haz Class: 9; Packing Group: III Crystalline; Linear Formula: CuBr·S(CH3)2 ; MW: 205.58 Melting Point: 132° (decomposes) Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Crystalline

Formula: C₂H₆BrCuS
Melting Pt: 132 °C (dec.)
MDL Number: MFCD00043295
CAS Number: 54678-23-8
EINECS: 259-292-5
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR