α-Bromo-2-chlorophenylacetic acid ≥98%

Supplier: BeanTown Chemical

225195-25G 225195-5G
BT225195-25GEA 264.73 USD
BT225195-25G BT225195-5G
α-Bromo-2-chlorophenylacetic acid ≥98%
α-Bromo-2-chlorophenylacetic acid

CAS: 29270-30-2; MDL No: MFCD03428513 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C8H6BrClO2 ; MW: 249.49 Melting Point: 105-108°


  • Solid

Formula: C₈H₆BrClO₂
Melting Pt: 105…108 °C
MDL Number: MFCD03428513
CAS Number: 29270-30-2
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR