Bismuth(III) chloride, (max. 3% H₂O) ≥98% (dry weight)
Supplier: BeanTown Chemical
Synonyms:
Bismuth(III) chloride, Bismuth trichloride
225175-100G
225175-25G
225175-500G
BT225175-100GEA
134
USD
BT225175-100G
BT225175-25G
BT225175-500G
Bismuth(III) chloride, (max. 3% H₂O) ≥98% (dry weight)
Bismuth(III) chloride
CAS: 7787-60-2; EC No: 232-123-2; MDL No: MFCD00003461; RTECS: EB2690000
UN No: UN3260; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: BiCl3; MW: 315.34
Melting Point: 230-232°; Boiling Point: 447°
Hygroscopic
- RTECS: EB2690000
- Hygroscopic
- Solid
Formula:
BiCl₃ MW: 315.34 g/mol Boiling Pt: 447 °C Melting Pt: 230…232 °C |
MDL Number:
MFCD00003461 CAS Number: 7787-60-2 EINECS: 232-123-2 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...