1,1,2,2-Tetrachloroethane-[D₂] ≥99.5% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
1,1,2,2-Tetrachloroethane-[²H₂]
225105-25G
225105-5G
BT225105-25GEA
351.46
USD
BT225105-25G
BT225105-5G
1,1,2,2-Tetrachloroethane-[D₂] ≥99.5% (Isotopic)
1,1,2,2-Tetrachloroethane-[D₂]
CAS: 33685-54-0; EC No: 251-634-1; MDL No: MFCD00037672
UN No: UN1702; Haz Class: 6.1; Packing Group: II
Liquid; Linear Formula: Cl2CDCDCl2; MW: 169.86
Boiling Point: 145-146°/737 mmHg
Density (g/mL): 1.62; Refractive Index: 1.493
- Refractive index: 1.493
- Liquid
Formula:
Cl₂CDCDCl₂ MW: 169.83 g/mol Boiling Pt: 145…146 °C/737 mmHg Density: 1.62 g/mL |
MDL Number:
MFCD00037672 CAS Number: 33685-54-0 EINECS: 251-634-1 UN: 1702 ADR: 6.1,II |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...