Aluminium fluoride, anhydrous ≥99%

Supplier: BeanTown Chemical

224985-10G 224985-50G
BT224985-10GEA 83.98 USD
BT224985-10G BT224985-50G
Aluminium fluoride, anhydrous ≥99%
Aluminium fluoride

CAS: 7784-18-1; EC No: 232-051-1; MDL No: MFCD00003426 UN No: UN3260; Haz Class: 8; Packing Group: II Powder; Molecular Formula: AlF3; MW: 83.98 Melting Point: 1291° Density (g/mL): 3.1 Hygroscopic


  • Hygroscopic
  • Powder

Formula: AlF₃
Melting Pt: 1291 °C
Density: 3.1 g/mL
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR