3,4-Difluoroaniline ≥98%
Supplier: BeanTown Chemical
CAS: 3863-11-4; EC No: 223-381-7; MDL No: MFCD00007761; RTECS: CX9871900
UN No: UN2941 ; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H5F2N; MW: 129.11
Boiling Point: 77°/7 mmHg; Flash point: 85°C (185°F)
Density (g/mL): 1.302; Refractive Index: 1.513
- RTECS: CX9871900
- Refractive index: 1.513
- Liquid
Formula:
C₆H₅F₂N Boiling Pt: 77 °C/7 mmHg Density: 1.302 g/mL Flash Pt: 85 °C (185 °F) |
MDL Number:
MFCD00007761 CAS Number: 3863-11-4 EINECS: 223-381-7 UN: 2941 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...