Resorcinol ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,3-Dihydroxybenzene, 1,3-Benzenediol

224905-1KG 224905-250G 224905-50G
BT224905-1KGEA 172.05 USD
BT224905-1KG BT224905-250G BT224905-50G
Resorcinol ≥99%
Resorcinol

CAS: 108-46-3; EC No: 203-585-2; MDL No: MFCD00002269; RTECS: VG9625000 UN No: UN2876; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: C6H4(OH)2; MW: 110.11 Melting Point: 109-112°; Boiling Point: 280-281°; Flash point: 127°C (261°F) Density (g/mL): 1.272 Air Sensitive, Light Sensitive


  • RTECS: VG9625000
  • Air Sensitive, Light Sensitive
  • Solid

Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 280…281 °C
Melting Pt: 109…112 °C
Density: 1.272 g/mL
Flash Pt: 127 °C (261 °F)
MDL Number: MFCD00002269
CAS Number: 108-46-3
EINECS: 203-585-2
UN: 2876
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR