Manganese(III) fluoride ≥98%
Supplier: BeanTown Chemical
224870-10G
224870-50G
BT224870-10GEA
44
USD
BT224870-10G
BT224870-50G
Manganese(III) fluoride ≥98%
Manganese(III) fluoride
CAS: 7783-53-1; EC No: 232-006-6; MDL No: MFCD00016223
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Linear Formula: MnF3; MW: 111.93
Density (g/mL): 3.54
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
F₃Mn MW: 111.93 Density: 3.54 g/mL |
MDL Number:
MFCD00016223 CAS Number: 7783-53-1 EINECS: 232-006-6 UN: 3288 ADR: 6.1 III Merck Index: 12,05784 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...