Butyronitrile ≥99%
Supplier: BeanTown Chemical
Synonyms:
n-Butyronitrile
224865-1L
224865-250ML
BT224865-1LEA
62.97
USD
BT224865-1L
BT224865-250ML
Butyronitrile ≥99%
Butyronitrile
CAS: 109-74-0; EC No: 203-700-6; MDL No: MFCD00001968; RTECS: ET8750000
UN No: UN2411; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: CH3CH2CH2CN; Molecular Formula: C4H7N; MW: 69.11
Melting Point: -112°; Boiling Point: 115-117°; Flash point: 18°C (64°F)
Density (g/mL): 0.794; Refractive Index: 1.384
- RTECS: ET8750000
- Refractive index: 1.384
- Liquid
Formula:
C₄H₇N MW: 69.11 g/mol Boiling Pt: 115…117 °C Melting Pt: –112 °C Density: 0.794 g/mL Flash Pt: 18 °C (64 °F) |
MDL Number:
MFCD00001968 CAS Number: 109-74-0 EINECS: 203-700-6 UN: 2411 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...