Silver trifluoroacetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Silver(I) trifluoroacetate
224855-25G
224855-5G
BT224855-25GEA
158.63
USD
BT224855-25G
BT224855-5G
Silver trifluoroacetate ≥98%
Silver trifluoroacetate
CAS: 2966-50-9; EC No: 221-004-0; MDL No: MFCD00013199
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline/Powder; Linear Formula: CF3COOAg; MW: 220.88
Melting Point: 257-260° (decomposes)
Hygroscopic , Light Sensitive
- Hygroscopic, Light Sensitive
- Crystalline/Powder
Formula:
C₂AgF₃O₂ MW: 220.88 Melting Pt: 257…260 °C (dec.) |
MDL Number:
MFCD00013199 CAS Number: 2966-50-9 EINECS: 221-004-0 ADR: 9 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...