N,N-Dimethylethylamine ≥99%

Supplier: BeanTown Chemical

Synonyms: DMEA

224825-100ML 224825-500ML
BT224825-100MLEA 39.97 USD
BT224825-100ML BT224825-500ML
N,N-Dimethylethylamine ≥99%
N,N-Dimethylethylamine

CAS: 598-56-1; EC No: 209-940-8; MDL No: MFCD00009039 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: (CH3)2NCH2CH3; Molecular Formula: C4H11N; MW: 73.14 Melting Point: -140°; Boiling Point: 36-38°; Flash point: -36°C (-33°F) Density (g/mL): 0.675; Refractive Index: 1.372 Light Sensitive


  • Refractive index: 1.372
  • Light Sensitive
  • Liquid

Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 36…38 °C
Melting Pt: –140 °C
Density: 0.675 g/mL
Flash Pt: –36 °C (–33 °F)
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8
ADR: 3 (8),II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR