Potassium tetracyanoplatinate(II) trihydrate ≥99.9%

Supplier: BeanTown Chemical
Danger

224765-1G 224765-5G
BT224765-1GEA 159 USD
BT224765-1G BT224765-5G
Potassium tetracyanoplatinate(II) trihydrate ≥99.9%
Potassium tetracyanoplatinate(II) trihydrate

CAS: 14323-36-5; EC No: 209-236-0; MDL No: MFCD00149921 UN No: UN1588; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C4N4K2Pt·3H2O; MW: 431.39


  • Soluble in hot water

Formula: C₄K₂N₄Pt·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149921
CAS Number: 38245-27-1
EINECS: 209-236-0
UN: 1588
ADR: 6.1,III
Merck Index: 14,7685

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR