2-Chloroethylammonium chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

224760-1KG 224760-250G 224760-50G
BT224760-1KGEA 363 USD
BT224760-1KG BT224760-250G BT224760-50G
2-Chloroethylammonium chloride ≥98%
2-Chloroethylammonium chloride

CAS: 870-24-6; EC No: 212-793-2; MDL No: MFCD00012887; RTECS: KR1800000 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: ClCH2CH2NH2·HCl; Molecular Formula: C2H6ClN·HCl; MW: 115.99 Melting Point: 140-150° Hygroscopic


  • RTECS: KR1800000
  • Hygroscopic
  • Crystalline

Formula: C₂H₇Cl₂N
MW: 115.99 g/mol
Melting Pt: 140…150 °C
MDL Number: MFCD00012887
CAS Number: 870-24-6
EINECS: 212-793-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR