Potassium fluoride ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

224685-25G 224685-5G
BT224685-25GEA 194 USD
BT224685-25G BT224685-5G
Potassium fluoride ≥99.9% (trace metals basis)
Potassium fluoride

CAS: 7789-23-3; EC No: 232-151-5; MDL No: MFCD00011398; RTECS: TT0700000 UN No: UN1812; Haz Class: 6.1; Packing Group: III Powder/Chunks; Linear Formula: KF; MW: 58.10 Melting Point: 858°; Boiling Point: 1505° Density (g/mL): 2.48; Refractive Index: 1.363 Hygroscopic


  • RTECS: TT0700000
  • Refractive index: 1.363
  • Hygroscopic
  • Powder/Chunks

Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505 °C
Melting Pt: 858 °C
Density: 2.48 g/mL
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR