3-Fluoroaniline ≥98%

Supplier: BeanTown Chemical

224665-100G 224665-25G 224665-500G
BT224665-100GEA 178 USD
BT224665-100G BT224665-25G BT224665-500G
3-Fluoroaniline ≥98%
3-Fluoroaniline

CAS: 372-19-0; EC No: 206-747-0; MDL No: MFCD00007758; RTECS: BY1400000 UN No: UN2941; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H6FN; MW: 111.12 Flash point: 77°C (171°F) Density (g/mL): 1.156; Refractive Index: 1.542


  • RTECS: BY1400000
  • Refractive index: 1.542
  • Liquid

Formula: C₆H₆FN
Density: 1.156 g/mL
Flash Pt: 77 °C (171 °F)
MDL Number: MFCD00007758
CAS Number: 372-19-0
EINECS: 206-747-0
UN: 2941
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR