Copper(II) acetate, anhydrous

Supplier: BeanTown Chemical

224655-100G 224655-25G 224655-500G
BT224655-100GEA 175.91 USD
BT224655-100G BT224655-25G BT224655-500G
Copper(II) acetate, anhydrous
Copper(II) acetate

CAS: 142-71-2; EC No: 205-553-3; MDL No: MFCD00008690; RTECS: AG3480000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: Cu(OOCCH3)2; MW: 181.63 Hygroscopic


  • RTECS: AG3480000
  • Hygroscopic
  • Powder

Formula: CuCH₃CO₂
MDL Number: MFCD00008690
CAS Number: 142-71-2
EINECS: 205-553-3
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR