1,4-Diaminobutane ≥98%

Supplier: BeanTown Chemical

Synonyms: Putrescine, 1,4-Butanediamine, Tetramethylendiamine

224575-100G 224575-25G 224575-500G
BT224575-100GEA 86.38 USD
BT224575-100G BT224575-25G BT224575-500G
1,4-Diaminobutane ≥98%
1,4-Diaminobutane

CAS: 110-60-1; EC No: 203-782-3; MDL No: MFCD00008235; RTECS: EJ6800000 UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II Liquid/Low Melting Solid; Molecular Formula: C4H12N2; MW: 88.15 Melting Point: 25-28°; Boiling Point: 158-160°; Flash point: 51°C (124°F) Density (g/mL): 0.877; Refractive Index: 1.457 Air Sensitive , Hygroscopic


  • RTECS: EJ6800000
  • Refractive index: 1.457
  • Air Sensitive, Hygroscopic
  • Liquid/Low Melting Solid

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 158…160 °C
Melting Pt: 25…28 °C
Density: 0.877 g/mL
Flash Pt: 51 °C (124 °F)
MDL Number: MFCD00008235
CAS Number: 110-60-1
EINECS: 203-782-3
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR