Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥94%

Supplier: BeanTown Chemical

Synonyms: Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate

224530-25G 224530-5G
BT224530-25GEA 180 USD
BT224530-25G BT224530-5G
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate ≥94%
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate

CAS: 363-58-6; MDL No: MFCD00041540 UN No: UN1760; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C6H6ClF3O3; MW: 218.56 Boiling Point: 67-71°/35 mmHg; Flash point: 67°C (153°F) Density (g/mL): 1.39


  • Liquid

Formula: C₆H₆ClF₃O₃
MW: 218.556 g/mol
Boiling Pt: 67…71 °C/35 mmHg
Density: 1.39 g/mL
Flash Pt: 67 °C (153 °F)
MDL Number: MFCD00041540
CAS Number: 363-58-6
ADR: 8,III

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR