3-(Trifluoromethyl)-p-anisidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-(Trifluoromethyl)-4-anisidine, 5-Amino-2-methoxybenzotrifluoride, 4-Methoxy-3-(trifluoromethyl)aniline, 3-(Trifluoromethyl)-4-methoxyaniline, 4-Amino-2-(trifluoromethyl)anisole

224510-1G 224510-5G
BT224510-1GEA 42 USD
BT224510-1G BT224510-5G
3-(Trifluoromethyl)-p-anisidine ≥98%
3-(Trifluoromethyl)-p-anisidine

CAS: 393-15-7; MDL No: MFCD00043460 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C8H8F3NO ; MW: 191.15 Melting Point: 57-61°; Boiling Point: 112°/18 mmHg


  • Crystalline/Powder

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 112 °C/18 mmHg
Melting Pt: 57…61 °C
MDL Number: MFCD00043460
CAS Number: 393-15-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR