Cetrimonium bromide ≥98%

Supplier: BeanTown Chemical

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

224475-100G 224475-500G
BT224475-100GEA 53.3 USD
BT224475-100G BT224475-500G
Cetrimonium bromide ≥98%
Cetrimonium bromide

CAS: 57-09-0; EC No: 200-311-3; MDL No: MFCD00011772; RTECS: BQ7875000 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Linear Formula: CH3(CH2)15N(Br)(CH3)3; Molecular Formula: C19H42BrN; MW: 364.45 Melting Point: 248-251°; Flash point: 244°C (471°F) Hygroscopic


  • RTECS: BQ7875000
  • Hygroscopic
  • Solid

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Melting Pt: 248…251 °C
Flash Pt: 244 °C (471 °F)
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR