4-Bromobenzenesulphonyl chloride ≥98%

Supplier: BeanTown Chemical

224380-100G 224380-25G 224380-500G
BT224380-100GEA 153 USD
BT224380-100G BT224380-25G BT224380-500G
4-Bromobenzenesulphonyl chloride ≥98%
4-Bromobenzenesulphonyl chloride

CAS: 98-58-8; EC No: 202-683-2; MDL No: MFCD00007437 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline/Powder; Molecular Formula: C6H4BrClO2S; MW: 255.52 Melting Point: 73-75°; Boiling Point: 153°/15 mmHg Moisture Sensitive


  • Moisture Sensitive
  • Crystalline/Powder

Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 287
Melting Pt: 74
MDL Number: MFCD00007437
CAS Number: 98-58-8
EINECS: 202-683-2
UN: 3261
ADR: 8 II
Merck Index: 13,01391

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR