Copper(II) sulfate, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
Cupric sulphate
224330-1KG
224330-250G
224330-5KG
BT224330-1KGEA
234.74
USD
BT224330-1KG
BT224330-250G
BT224330-5KG
Copper(II) sulfate, anhydrous ≥98%
Copper(II) sulphate
CAS: 7758-98-7; EC No: 231-847-6; MDL No: MFCD00010981; RTECS: GL8800000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: CuSO4; MW: 159.60
Melting Point: 560° (decomposes)
Density (g/mL): 3.603
Hygroscopic
- RTECS: GL8800000
- Hygroscopic
- Powder
Formula:
CuSO₄ MW: 159.61 g/mol Melting Pt: 560 °C (dec.) Density: 3.603 g/mL |
MDL Number:
MFCD00010981 CAS Number: 7758-98-7 EINECS: 231-847-6 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...