Mercury(II) telluride ≥99%

Supplier: BeanTown Chemical

Synonyms: Mercurous telluride

224275-25G 224275-5G
BT224275-25GEA 963 USD
BT224275-25G BT224275-5G
Mercury(II) telluride ≥99%
Mercury(II) telluride

CAS: 12068-90-5; EC No: 235-108-9; MDL No: MFCD00016133 UN No: UN2025; Haz Class: 6.1; Packing Group: II -100 Mesh Powder; Molecular Formula: HgTe; MW: 328.19 Melting Point: 673° Density (g/mL): 8.63


  • -100 Mesh Powder

Formula: HgTe
MW: 328.19
Melting Pt: 550
Density: 8.63 g/mL
MDL Number: MFCD00016133
CAS Number: 12068-90-5
EINECS: 235-108-9
UN: 2025
ADR: 6.1 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR