(±)-3,3-Dimethyl-2-butanol ≥99%
Supplier: BeanTown Chemical
Synonyms:
Pinacolyl alcohol
224265-10G
224265-50G
BT224265-10GEA
33
USD
BT224265-10G
BT224265-50G
(±)-3,3-Dimethyl-2-butanol ≥99%
(±)-3,3-Dimethyl-2-butanol
CAS: 464-07-3; EC No: 207-347-9; MDL No: MFCD00004522; RTECS: EL2276000
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3)3CCH(OH)CH3; Molecular Formula: C6H14O; MW: 102.17
Melting Point: 4.8°; Boiling Point: 119-121°; Flash point: 26°C (79°F)
Density (g/mL): 0.812; Refractive Index: 1.415
- RTECS: EL2276000
- Refractive index: 1.415
- Liquid
Formula:
C₆H₁₄O MW: 102.18 g/mol Boiling Pt: 119…121 °C Melting Pt: 4.8 °C Density: 0.812 g/mL Flash Pt: 26 °C (79 °F) |
MDL Number:
MFCD00004522 CAS Number: 464-07-3 EINECS: 207-347-9 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...