(R)-(+)-1,2-Epoxypropane ≥99%
Supplier: BeanTown Chemical
Synonyms:
(R)-(+)-Propene oxide, (R)-(+)-Methyloxirane, (R)-(+)-1,2-Propylene oxide, (R)-(+)-Propylene oxide
224260-100G
224260-25G
224260-5G
BT224260-100GEA
792
USD
BT224260-100G
BT224260-25G
BT224260-5G
(R)-(+)-1,2-Epoxypropane ≥99%
(R)-(+)-1,2-Epoxypropane
CAS: 15448-47-2; MDL No: MFCD00066211; RTECS: UJ2650000
UN No: UN1280; Haz Class: 3; Packing Group: I
Liquid; Molecular Formula: C3H6O; MW: 58.08
Boiling Point: 33-34°; Flash point: -37°C (-35°F)
Density (g/mL): 0.829; Refractive Index: 1.366; Optical Rotation: [α]20/D +14°, neat
- RTECS: UJ2650000
- Optical rotation: [α]20/D +14°, neat
- Refractive index: 1.366
- Liquid
Formula:
C₃H₆O MW: 58.08 g/mol Boiling Pt: 33…34 °C Density: 0.829 g/mL Flash Pt: –37 °C (–35 °F) |
MDL Number:
MFCD00066211 CAS Number: 15448-47-2 UN: 1280 ADR: 3,I |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...