Choline hydroxide 46% (w/w) in water
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxyethyltrimethylammonium Hydroxide, Choline
224210-100ML
224210-25ML
224210-500ML
BT224210-100MLEA
80.25
USD
BT224210-100ML
BT224210-25ML
BT224210-500ML
Choline hydroxide 46% (w/w) in water
Choline hydroxide
CAS: 123-41-1; EC No: 204-625-1; MDL No: MFCD00002831; RTECS: GA4025500
UN No: UN3267; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: HOCH2CH2N(CH3)3OH; Molecular Formula: C5H16NO2; MW: 121.18
Density (g/mL): 1.073; Refractive Index: 1.430
- RTECS: GA4025500
- Refractive index: 1.430
- Liquid
Formula:
C₅H₁₅NO₂ MW: 121.18 g/mol Density: 1.073 g/mL |
MDL Number:
MFCD00002831 CAS Number: 123-41-1 EINECS: 204-625-1 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...