p-Toluenesulfonyl hydrazide ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Methylbenzenesulphonylhydrazide, 4-Toluenesulphonyl hydrazide, 4-Methylbenzenesulphonhydrazide, 4-Methylbenzenesulphonic acid hydrazide, 4-Methylbenzenesulphonohydrazide, Toluene-4-sulphonohydrazide
224155-100G
224155-25G
224155-500G
BT224155-100GEA
59.45
USD
BT224155-100G
BT224155-25G
BT224155-500G
p-Toluenesulfonyl hydrazide ≥98%
p-Toluenesulphonyl hydrazide
CAS: 1576-35-8; EC No: 216-407-3; MDL No: MFCD00007588; RTECS: MW0210000
UN No: UN3226; Haz Class: 4.1; Packing Group: II
Crystalline; Linear Formula: CH3C6H4SO2HNH2; Molecular Formula: C7H10N2O2S; MW: 186.23
Melting Point: 103-108°; Flash point: 60°C (140°F)
Density (g/mL): 1.4
- RTECS: MW0210000
- Crystalline
Formula:
C₇H₁₀N₂O₂S MW: 186.23 g/mol Melting Pt: 103…108 °C Density: 1.4 g/mL Flash Pt: 60 °C (140 °F) |
MDL Number:
MFCD00007588 CAS Number: 1576-35-8 EINECS: 216-407-3 ADR: 4.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...