(p-Toluenesulfonyl)methyl isocyanide ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide

224035-100G 224035-25G 224035-5G
BT224035-100GEA 517 USD
BT224035-100G BT224035-25G BT224035-5G
(p-Toluenesulfonyl)methyl isocyanide ≥97%
(p-Toluenesulphonyl)methyl isocyanide

CAS: 36635-61-7; EC No: 253-140-1; MDL No: MFCD00000005 UN No: UN2811; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: CH3C6H4SO2CH2NC; Molecular Formula: C9H9NO2S; MW: 195.24 Melting Point: 109-113° Air Sensitive, Light Sensitive, Moisture Sensitive


  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Crystalline

Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 109…113 °C
MDL Number: MFCD00000005
CAS Number: 36635-61-7
EINECS: 253-140-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR