tert-Butyl chloroacetate ≥98%
Supplier: BeanTown Chemical
CAS: 107-59-5; EC No: 203-506-1; MDL No: MFCD00000930; RTECS: AF8985500
UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II
Liquid; Linear Formula: ClCH2COOC(CH3)3; Molecular Formula: C6H11ClO2; MW: 150.61
Boiling Point: 48-49°/11mm Hg; Flash point: 47°C (117°F)
Density (g/mL): 1.053; Refractive Index: 1.423
Moisture Sensitive
- RTECS: AF8985500
- Refractive index: 1.423
- Moisture Sensitive
- Liquid
Formula:
C₆H₁₁ClO₂ Boiling Pt: 48…49 °C/11mm Hg Density: 1.053 g/mL Flash Pt: 47 °C (117 °F) |
MDL Number:
MFCD00000930 CAS Number: 107-59-5 EINECS: 203-506-1 UN: 2929 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...