Lithium perchlorate trihydrate
Supplier: BeanTown Chemical
223960-1KG
223960-250G
BT223960-1KGEA
164
USD
BT223960-1KG
BT223960-250G
Lithium perchlorate trihydrate
Lithium perchlorate trihydrate
CAS: 13453-78-6; EC No: 232-237-2; MDL No: MFCD00149765; RTECS: SC8750000
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: LiClO4·3H2O; MW: 160.45
Density (g/mL): 1.84
- RTECS: SC8750000
- Crystalline
Formula:
ClLiO₄·3H₂O MW: 160.44 Melting Pt: 91.85 Density: 1.84 g/mL |
MDL Number:
MFCD00149765 CAS Number: 13453-78-6 EINECS: 232-237-2 UN: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...