Copper(II) oxide
Supplier: BeanTown Chemical
Synonyms:
Cupric oxide, Copper oxide, Copper monoxide granulated
223895-10G
223895-50G
BT223895-10GEA
38
USD
BT223895-10G
BT223895-50G
Copper(II) oxide
Copper(II) oxide
CAS: 1317-38-0; EC No: 215-269-1; MDL No: MFCD00010979; RTECS: GL7900000
UN No: UN3077; Haz Class: 9; Packing Group: III
30-50nm APS Powder, S.A. 13 m2/g; Molecular Formula: CuO; MW: 79.55
Melting Point: 1330°
Density (g/mL): 6.4
- RTECS: GL7900000
- 30-50nm APS Powder, S.A. 13 m²/g
Formula:
CuO MW: 79.55 g/mol Boiling Pt: 2000 Melting Pt: 1326 Density: 6.4 g/mL |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9 III Merck Index: 13,02674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...