Chromium(III) nitrate nonahydrate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate
223885-100G
223885-25G
223885-5G
BT223885-100GEA
534
USD
BT223885-100G
BT223885-25G
BT223885-5G
Chromium(III) nitrate nonahydrate ≥99.99% (trace metals basis)
Chromium(III) nitrate nonahydrate
CAS: 7789-02-8; EC No: 236-921-1; MDL No: MFCD00149659; RTECS: GB6300000
UN No: UN2720; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Cr(NO3)3·9H2O; MW: 400.14
Melting Point: 60°
Density (g/mL): 1.8
- RTECS: GB6300000
- Crystalline
Formula:
CrN₃O₉·9H₂O MW: 400.15 g/mol Melting Pt: 60 °C Density: 1.8 g/mL |
MDL Number:
MFCD00149659 CAS Number: 7789-02-8 EINECS: 236-921-1 UN: 2720 ADR: 5.1,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...