2-Chloronicotinonitrile ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile
223870-1G
223870-25G
223870-5G
BT223870-1GEA
21
USD
BT223870-1G
BT223870-25G
BT223870-5G
2-Chloronicotinonitrile ≥98%
2-Chloronicotinonitrile
CAS: 6602-54-6; EC No: 229-550-1; MDL No: MFCD00014628
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H3ClN2; MW: 138.55
Melting Point: 104-107°
- Solid
Formula:
C₆H₃ClN₂ MW: 138.56 g/mol Melting Pt: 104…107 °C |
MDL Number:
MFCD00014628 CAS Number: 6602-54-6 EINECS: 229-550-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...