2,4,4-Trimethyl-2-pentene ≥97%

Supplier: BeanTown Chemical

223860-100G 223860-25G
BT223860-100GEA 298 USD
BT223860-100G BT223860-25G
2,4,4-Trimethyl-2-pentene ≥97%
2,4,4-Trimethyl-2-pentene

CAS: 107-40-4; EC No: 203-488-5; MDL No: MFCD00008902 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CCH=C(CH3)2; Molecular Formula: C8H16; MW: 112.21 Melting Point: -106°; Boiling Point: 104°; Flash point: 2°C (36°F) Density (g/mL): 0.72; Refractive Index: 1.416


  • Refractive index: 1.416
  • Liquid

Formula: C₈H₁₆
Boiling Pt: 104 °C
Melting Pt: –106 °C
Density: 0.72 g/mL
Flash Pt: 2 °C (36 °F)
MDL Number: MFCD00008902
CAS Number: 107-40-4
EINECS: 203-488-5
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR