(1R)-(-)-Camphor-10-sulphonyl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: (1R)-(-)-2-Oxobornane-10-sulphonyl chloride, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonyl chloride, (1R)-(-)-Camphor-10-sulphonic acid chloride, L(-)-Camphor-10-sulphonyl chloride

223850-1G 223850-25G 223850-5G
BT223850-1GEA 24 USD
BT223850-1G BT223850-25G BT223850-5G
(1R)-(-)-Camphor-10-sulphonyl chloride ≥97%
(1R)-(-)-Camphor-10-sulphonyl chloride

CAS: 39262-22-1; MDL No: MFCD00064155 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C10H15ClO3S ; MW: 250.74 Melting Point: 66-68° Optical Rotation: [α]18/D −33°, c = 3 in chloroform Moisture Sensitive


  • Optical rotation: [α]18/D −33°, c = 3 in chloroform
  • Moisture Sensitive
  • Crystalline

Formula: C₁₀H₁₅ClO₃S
Melting Pt: 66…68 °C
MDL Number: MFCD00064155
CAS Number: 39262-22-1
UN: 3261
ADR: 8,I

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR