Ethanolamine ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: 2-Aminoethanol, Monoethanolamine

223825-4X500ML 223825-500ML
BT223825-4X500EA 139.86 USD
BT223825-4X500 BT223825-500ML
Ethanolamine ≥99.0% ACS
Ethanolamine

CAS: 141-43-5; EC No: 205-483-3; MDL No: MFCD00008183; RTECS: KJ5775000 UN No: UN2491; Haz Class: 8; Packing Group: III Liquid; Linear Formula: HOCH2CH2NH2; Molecular Formula: C2H7NO; MW: 61.08 Melting Point: 10-11°; Boiling Point: 170°; Flash point: 86°C (187°F) Density (g/mL): 1.012; Refractive Index: 1.454 Air Sensitive , Hygroscopic


  • RTECS: KJ5775000
  • Refractive index: 1.454
  • Air Sensitive, Hygroscopic
  • Liquid

Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 170 °C
Melting Pt: 10…11 °C
Density: 1.012 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR