1-(4-Fluorophenyl)piperazine dihydrochloride ≥98%

Supplier: BeanTown Chemical

223810-100G 223810-25G 223810-500G
BT223810-100GEA 112 USD
BT223810-100G BT223810-25G BT223810-500G
1-(4-Fluorophenyl)piperazine dihydrochloride ≥98%
1-(4-Fluorophenyl)piperazine dihydrochloride

CAS: 64090-19-3; MDL No: MFCD00012765 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C10H13FN2·2HCl; MW: 253.15


  • Crystalline/Powder

Formula: C₁₀H₁₅Cl₂FN₂
MW: 253.15 g/mol
MDL Number: MFCD00012765
CAS Number: 64090-19-3
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR